(4-Chloro-2-ethoxyphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

319985-5G 319985-500MG 319985-25G 319985-1G
77632-688EA 1441 USD
77632-688 77652-640 77654-984 77642-002
(4-Chloro-2-ethoxyphenyl)(cyclopropyl)methanol ≥95%
(4-Chloro-2-ethoxyphenyl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD35176610
CAS Number: 3002501-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR