1-(4-Phenoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

319876-5G 319876-25G 319876-1G 319876-500MG
77632-684EA 1171 USD
77632-684 77654-982 77642-000 77650-040
1-(4-Phenoxyphenyl)propan-1-one ≥95%
1-(4-Phenoxyphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 226.27 g/mol
MW: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11126939
CAS Number: 889-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR