2-(4-Phenoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

319874-5G 319874-25G 319874-1G 319874-500MG
77632-682EA 1186 USD
77632-682 77658-692 77646-014 77646-016
2-(4-Phenoxyphenyl)propan-2-ol ≥95%
2-(4-Phenoxyphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 228.286 g/mol
MW: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD20528503
CAS Number: 4974-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR