Methyl(4-phenoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

319872-5G 319872-25G 319872-1G 319872-500MG
77632-680EA 1186 USD
77632-680 77658-690 77646-012 77650-038
Methyl(4-phenoxyphenyl)sulfane ≥95%
Methyl(4-phenoxyphenyl)sulfane

Precaución: For Research Use Only

Formula: C₁₃H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD11617758
CAS Number: 21134-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR