2-Bromo-4-(tert-butyl)-1-((3-methylbenzyl)oxy)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
319849-5G
319849-1G
319849-25G
319849-500MG
77628-568EA
1484.23
USD
77628-568
77638-028
77658-688
77646-010
2-Bromo-4-(tert-butyl)-1-((3-methylbenzyl)oxy)benzene ≥97%
2-Bromo-4-(tert-butyl)-1-((3-methylbenzyl)oxy)benzene
Precaución: For Research Use Only
Formula:
333.2627 g/mol MW: C₁₈H₂₁BrO Storage Temperature: Ambient |
MDL Number:
MFCD35176499 CAS Number: 3002454-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...