2-Bromo-4-(tert-butyl)-1-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

319841-5G 319841-25G 319841-1G 319841-500MG
77628-566EA 1484.23 USD
77628-566 77654-980 77641-998 77646-008
2-Bromo-4-(tert-butyl)-1-propoxybenzene ≥97%

Precaución: For Research Use Only

Formula: C₁₃H₁₉BrO
Storage Temperature: Ambient
MDL Number: MFCD35176492
CAS Number: 3002524-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR