6-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%

Supplier: AOBChem USA

Warning

319728-5G 319728-25G 319728-1G 319728-500MG
77632-678EA 855 USD
77632-678 77658-686 77641-996 77650-036
6-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%
6-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-amine

Precaución: For Research Use Only

Formula: 260.0351 g/mol
MW: C₆H₅IN₄
Storage Temperature: Ambient
CAS Number: 1245647-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR