2-Benzyl-4-bromoaniline ≥95%

Supplier: AOBChem USA

Warning

319643-5G 319643-1G 319643-500MG
77658-684EA 1811 USD
77658-684 77641-994 77646-006
2-Benzyl-4-bromoaniline ≥95%
2-Benzyl-4-bromoaniline

Precaución: For Research Use Only

Formula: 262.1450 g/mol
MW: C₁₃H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD12923305
CAS Number: 86233-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR