2,3,5-Trimethylthiophene ≥97%

Supplier: AOBChem USA

Warning

319489-5G 319489-1G 319489-25G
77634-750EA 240 USD
77634-750 77638-026 77658-682
2,3,5-Trimethylthiophene ≥97%
2,3,5-Trimethylthiophene

Precaución: For Research Use Only

Formula: C₇H₁₀S
Storage Temperature: Ambient
MDL Number: MFCD00142784
CAS Number: 1795-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR