4-Methoxy-α,α-dimethyl-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

319488-5G 319488-1G 319488-500MG
77628-564EA 1484.23 USD
77628-564 77638-024 77646-004
4-Methoxy-α,α-dimethyl-1-naphthalenemethanol ≥95%
4-Methoxy-α,α-dimethyl-1-naphthalenemethanol

Precaución: For Research Use Only

Formula: 216.28 g/mol
MW: C₁₄H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD35176277
CAS Number: 107619-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR