2,6-Difluorobenzene-1,4-diol ≥95%

Supplier: AOBChem USA

Warning

31933-5G 31933-25G 31933-1G 31933-500MG
77632-676EA 812 USD
77632-676 77654-976 77641-992 77650-034
2,6-Difluorobenzene-1,4-diol ≥95%
2,6-Difluorobenzene-1,4-diol

Precaución: For Research Use Only

Formula: C₆H₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18394832
CAS Number: 84959-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR