4-[(tert-Butyldimethylsilyl)oxy]benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

318919-5G 318919-25G
77634-748EA 59 USD
77634-748 77658-680
4-[(tert-Butyldimethylsilyl)oxy]benzaldehyde ≥97%
4-[(tert-Butyldimethylsilyl)oxy]benzaldehyde

Precaución: For Research Use Only

Formula: C₁₃H₂₀O₂Si
Storage Temperature: Ambient
MDL Number: MFCD05865174
CAS Number: 120743-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR