1-[6-(Cyclohexyloxy)-3-pyridinyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

318760-5G 318760-25G 318760-1G 318760-500MG
77632-674EA 1381 USD
77632-674 77658-678 77641-990 77650-032
1-[6-(Cyclohexyloxy)-3-pyridinyl]ethanone ≥95%
1-[6-(Cyclohexyloxy)-3-pyridinyl]ethanone

Precaución: For Research Use Only

Formula: 219.28 g/mol
MW: C₁₃H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD24260559
CAS Number: 1545051-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR