2-(Cyclohexyloxy)-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

318738-5G 318738-1G 318738-25G 318738-500MG
77632-672EA 1055 USD
77632-672 77638-022 77654-974 77650-030
2-(Cyclohexyloxy)-4-iodopyridine ≥97%
2-(Cyclohexyloxy)-4-iodopyridine

Precaución: For Research Use Only

Formula: C₁₁H₁₄INO
Storage Temperature: Ambient
MDL Number: MFCD35176066
CAS Number: 1430753-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR