6-(Cyclohexyloxy)-3-pyridinecarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

318718-5G 318718-1G 318718-500MG
77654-972EA 2722 USD
77654-972 77641-988 77650-028
6-(Cyclohexyloxy)-3-pyridinecarboxylic acid ≥95%
6-(Cyclohexyloxy)-3-pyridinecarboxylic acid

Precaución: For Research Use Only

Formula: 221.25 g/mol
MW: C₁₂H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD09258837
CAS Number: 509101-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR