(2-Bromo-6-fluoro-3-propoxyphenyl)(cyclopropyl)methanol ≥97%

Supplier: AOBChem USA

Warning

318585-5G 318585-25G 318585-1G 318585-500MG
77632-666EA 1441 USD
77632-666 77654-970 77641-984 77650-022
(2-Bromo-6-fluoro-3-propoxyphenyl)(cyclopropyl)methanol ≥97%
(2-Bromo-6-fluoro-3-propoxyphenyl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: 303.1673 g/mol
MW: C₁₃H₁₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD35176012
CAS Number: 3002531-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR