4,5-Difluorosalicylaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Hydroxy-4,5-difluorobenzaldehyde, 4,5-Difluoro-2-hydroxybenzaldehyde

31840-5G 31840-25G
77634-746EA 128 USD
77634-746 77654-968
4,5-Difluorosalicylaldehyde ≥97%
4,5-Difluorosalicylaldehyde

Precaución: For Research Use Only

Formula: C₇H₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08458005
CAS Number: 199287-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR