2-(Difluoromethoxy)-4-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

31806-1G 31806-5G 31806-500MG
77638-012EA 580 USD
77638-012 77658-668 77645-994
2-(Difluoromethoxy)-4-fluorobenzaldehyde ≥95%
2-(Difluoromethoxy)-4-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: 190.119 g/mol
MW: C₈H₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD14698500
CAS Number: 1214333-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR