2-Chloro-4-(2-propen-1-yloxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

317780-5G 317780-1G 317780-500MG
77654-966EA 2211 USD
77654-966 77641-982 77650-018
2-Chloro-4-(2-propen-1-yloxy)pyridine ≥97%
2-Chloro-4-(2-propen-1-yloxy)pyridine

Precaución: For Research Use Only

Formula: 169.61 g/mol
MW: C₈H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD18885843
CAS Number: 1340215-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR