2,6-Difluoro-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

31751-5G 31751-1G 31751-25G 31751-500MG
77632-664EA 656 USD
77632-664 77638-010 77658-664 77645-988
2,6-Difluoro-4-isopropoxybenzoic acid ≥97%
2,6-Difluoro-4-isopropoxybenzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30344673
CAS Number: 1993479-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR