1-Bromo-2-ethoxy-4-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

31736-5G 31736-1G 31736-500MG 31736-25G
77634-742EA 361 USD
77634-742 77638-008 77652-638 77658-662
1-Bromo-2-ethoxy-4-(trifluoromethoxy)benzene ≥95%
1-Bromo-2-ethoxy-4-(trifluoromethoxy)benzene

Precaución: For Research Use Only

Formula: 285.058 g/mol
MW: C₉H₈BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18917879
CAS Number: 1162261-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR