2,4-Difluoro-5-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31728-5G 31728-1G 31728-25G 31728-500MG
77632-662EA 1098 USD
77632-662 77638-004 77658-660 77650-014
2,4-Difluoro-5-methoxybenzyl alcohol ≥97%
2,4-Difluoro-5-methoxybenzyl alcohol

Precaución: For Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28401420
CAS Number: 1266114-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR