Cyclohexyl(2-fluoro-5-methylphenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

317056-5G 317056-1G 317056-500MG
77654-962EA 1898 USD
77654-962 77641-978 77645-984
Cyclohexyl(2-fluoro-5-methylphenyl)methanone ≥97%
Cyclohexyl(2-fluoro-5-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 220.28 g/mol
MW: C₁₄H₁₇FO
Storage Temperature: Ambient
MDL Number: MFCD17222557
CAS Number: 1343094-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR