5-Bromo-2-fluoropyridine-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

31703-5G 31703-1G 31703-500MG
77658-658EA 2710 USD
77658-658 77641-976 77650-012
5-Bromo-2-fluoropyridine-4-carbaldehyde ≥97%
5-Bromo-2-fluoropyridine-4-carbaldehyde

Precaución: For Research Use Only

Formula: 203.997 g/mol
MW: C₆H₃BrFNO
Storage Temperature: Ambient
MDL Number: MFCD16606514
CAS Number: 1227588-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR