Cyclobutyl(2-fluoro-5-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

317026-5G 317026-25G 317026-1G 317026-500MG
77632-660EA 1388 USD
77632-660 77658-656 77641-974 77650-010
Cyclobutyl(2-fluoro-5-methylphenyl)methanone ≥95%
Cyclobutyl(2-fluoro-5-methylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD21117553
CAS Number: 1498201-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR