(2-Fluoro-5-methylphenyl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

317012-1G 317012-5G 317012-500MG
77638-002EA 559 USD
77638-002 77654-960 77650-008
(2-Fluoro-5-methylphenyl)phenylmethanone ≥95%
(2-Fluoro-5-methylphenyl)phenylmethanone

Precaución: For Research Use Only

Formula: 214.24 g/mol
MW: C₁₄H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD16783206
CAS Number: 59396-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR