1,5-Dichloro-2-ethyl-3-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

316820-5G 316820-25G 316820-1G 316820-500MG
77628-554EA 1484.23 USD
77628-554 77654-956 77641-972 77645-982
1,5-Dichloro-2-ethyl-3-fluorobenzene ≥95%
1,5-Dichloro-2-ethyl-3-fluorobenzene

Precaución: For Research Use Only

Formula: 193.05 g/mol
MW: C₈H₇Cl₂F
Storage Temperature: Ambient
CAS Number: 1805127-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR