(E)-ethyl 4,4,4-trifluoro-2-(2-hydroxybenzylidene)-3-oxobutanoate ≥95%
Supplier: AOBChem USA
|
Warning
|
316718-5G
316718-1G
316718-25G
316718-500MG
77628-552EA
1484.23
USD
77628-552
77638-000
77658-654
77650-006
(E)-ethyl 4,4,4-trifluoro-2-(2-hydroxybenzylidene)-3-oxobutanoate ≥95%
(E)-ethyl 4,4,4-trifluoro-2-(2-hydroxybenzylidene)-3-oxobutanoate
Precaución: For Research Use Only
Formula:
C₁₃H₁₁F₃O₄ Storage Temperature: Ambient |
MDL Number:
MFCD35175082 CAS Number: 3002983-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...