2-Methoxy-3-(trifluoromethyl)isonicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

316651-1G 316651-5G 316651-500MG
77634-738EA 1118 USD
77634-738 77654-952 77650-004
2-Methoxy-3-(trifluoromethyl)isonicotinaldehyde ≥95%
2-Methoxy-3-(trifluoromethyl)isonicotinaldehyde

Precaución: For Research Use Only

Formula: 205.1339 g/mol
MW: C₈H₆F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD16611535
CAS Number: 1227572-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR