2-(4-(Benzyloxy)-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

31654-5G 31654-25G 31654-1G 31654-500MG
77632-658EA 1380 USD
77632-658 77658-652 77641-970 77645-978
2-(4-(Benzyloxy)-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-(Benzyloxy)-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For Research Use Only

Formula: C₂₁H₂₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD18383422
CAS Number: 1257078-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR