Phenol, 2-methoxy-5-methyl-, 1-acetate ≥97%

Supplier: AOBChem USA

Warning

316452-5G 316452-25G 316452-1G 316452-500MG
77628-548EA 1484.23 USD
77628-548 77635-632 77641-968 77645-976
Phenol, 2-methoxy-5-methyl-, 1-acetate ≥97%
Phenol, 2-methoxy-5-methyl-, 1-acetate

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD31537697
CAS Number: 1688-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR