1-Fluoro-2-(propan-2-yloxy)-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

31625-5G 31625-1G 31625-25G
77634-734EA 243 USD
77634-734 77637-994 77654-948
1-Fluoro-2-(propan-2-yloxy)-4-(trifluoromethyl)benzene ≥97%
1-Fluoro-2-(propan-2-yloxy)-4-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 222.179 g/mol
MW: C₁₀H₁₀F₄O
Storage Temperature: Ambient
MDL Number: MFCD23113750
CAS Number: 1394936-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR