2-Bromo-3,5,6-trifluoro-4-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

315929-5G 315929-25G 315929-1G 315929-500MG
77628-546EA 1841.64 USD
77628-546 77658-646 77641-964 77645-974
2-Bromo-3,5,6-trifluoro-4-methoxybenzoic acid ≥95%
2-Bromo-3,5,6-trifluoro-4-methoxybenzoic acid

Precaución: For Research Use Only

Formula: C₈H₄BrF₃O₃
Storage Temperature: Ambient
CAS Number: 35844-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR