4-Bromo-N-methoxy-N-methylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

31567-5G 31567-25G 31567-1G 31567-500MG
77632-652EA 941 USD
77632-652 77654-944 77641-962 77645-972
4-Bromo-N-methoxy-N-methylpicolinamide ≥97%
4-Bromo-N-methoxy-N-methylpicolinamide

Precaución: For Research Use Only

Formula: C₈H₉BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13191447
CAS Number: 1005342-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR