Thiophene-2,5-dicarbaldehyde ≥97%

Supplier: AOBChem USA

Warning

31533-5G 31533-25G
77634-732EA 73 USD
77634-732 77654-942
Thiophene-2,5-dicarbaldehyde ≥97%
Thiophene-2,5-dicarbaldehyde

Precaución: For Research Use Only

Formula: C₆H₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD00216592
CAS Number: 932-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR