4'-Propyl[1,1'-biphenyl]-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

315303-5G 315303-1G 315303-25G 315303-500MG
77634-730EA 475 USD
77634-730 77637-992 77654-940 77649-998
4'-Propyl[1,1'-biphenyl]-4-carboxaldehyde ≥97%
4'-Propyl[1,1'-biphenyl]-4-carboxaldehyde

Precaución: For Research Use Only

Formula: 224.3 g/mol
MW: C₁₆H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD05981725
CAS Number: 93972-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR