3-Bromo-4-(2,2-difluoroethoxy)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

314296-5G 314296-25G 314296-1G 314296-500MG
77632-650EA 1092 USD
77632-650 77658-644 77641-960 77645-970
3-Bromo-4-(2,2-difluoroethoxy)benzonitrile ≥97%
3-Bromo-4-(2,2-difluoroethoxy)benzonitrile

Precaución: For Research Use Only

Formula: C₉H₆BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD24307538
CAS Number: 1553446-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR