1,2,3,5-Tetramethylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Isodurene

314290-5G 314290-25G
77634-728EA 59 USD
77634-728 77654-938
1,2,3,5-Tetramethylbenzene ≥97%
1,2,3,5-Tetramethylbenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008524
CAS Number: 527-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR