2-Fluoro-1-(4-methylnaphthalen-1-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

313135-5G 313135-1G 313135-25G 313135-500MG
77628-538EA 1484.23 USD
77628-538 77637-988 77654-934 77649-990
2-Fluoro-1-(4-methylnaphthalen-1-yl)ethanone ≥95%
2-Fluoro-1-(4-methylnaphthalen-1-yl)ethanone

Precaución: For Research Use Only

Formula: 202.2242 g/mol
MW: C₁₃H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD35173135
CAS Number: 3002436-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR