2-(4-Methylnaphthalen-1-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

313134-5G 313134-1G 313134-500MG
77628-536EA 1484.23 USD
77628-536 77637-986 77645-964
2-(4-Methylnaphthalen-1-yl)propan-2-ol ≥95%
2-(4-Methylnaphthalen-1-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 200.2762 g/mol
MW: C₁₄H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD09865153
CAS Number: 856206-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR