(6-Methoxybenzo[d][1,3]dioxol-5-yl)boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
312962-1G
312962-500MG
312962-5G
77634-726EA
812
USD
77634-726
77652-636
77658-636
(6-Methoxybenzo[d][1,3]dioxol-5-yl)boronic acid ≥97%
(6-Methoxybenzo[d][1,3]dioxol-5-yl)boronic acid
Precaución: For Research Use Only
Formula:
195.965 g/mol MW: C₈H₉BO₅ Storage Temperature: Ambient |
MDL Number:
MFCD22414926 CAS Number: 1432485-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...