6-Bromo-3-chloro-2-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

31261-5G 31261-25G 31261-1G 31261-500MG
77632-646EA 847 USD
77632-646 77658-632 77641-950 77645-958
6-Bromo-3-chloro-2-methoxybenzaldehyde ≥95%
6-Bromo-3-chloro-2-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: 249.489 g/mol
MW: C₈H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD32631801
CAS Number: 1628525-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR