4,6-Dibromo-2-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

31218-5G 31218-1G 31218-25G 31218-500MG
77634-720EA 298 USD
77634-720 77637-980 77654-928 77649-984
4,6-Dibromo-2-fluoro-3-methylbenzene ≥97%
4,6-Dibromo-2-fluoro-3-methylbenzene

Precaución: For Research Use Only

Formula: 283.92 g/mol
MW: C₇H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD31567220
CAS Number: 1357103-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR