Methyl 3,5-dichloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

31209-5G 31209-25G 31209-1G 31209-500MG
77632-644EA 786 USD
77632-644 77654-926 77641-946 77645-954
Methyl 3,5-dichloro-2-fluorobenzoate ≥97%
Methyl 3,5-dichloro-2-fluorobenzoate

Precaución: For Research Use Only

Formula: 223.029 g/mol
MW: C₈H₅Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD24108551
CAS Number: 1522125-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR