3,5-Dichloro-2-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31208-5G 31208-25G 31208-1G 31208-500MG
77632-642EA 1025 USD
77632-642 77654-924 77641-944 77645-952
3,5-Dichloro-2-fluorobenzyl alcohol ≥97%
3,5-Dichloro-2-fluorobenzyl alcohol

Precaución: For Research Use Only

Formula: C₇H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD28786034
CAS Number: 67640-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR