4-Benzyloxy-1-bromo-5-chloro-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

31205-5G 31205-1G 31205-25G 31205-500MG
77632-640EA 933 USD
77632-640 77637-978 77654-922 77649-982
4-Benzyloxy-1-bromo-5-chloro-2-fluorobenzene ≥97%
4-Benzyloxy-1-bromo-5-chloro-2-fluorobenzene

Precaución: For Research Use Only

Formula: 315.565 g/mol
MW: C₁₃H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD29088279
CAS Number: 1879026-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR