5-Chloro-2-fluoro-4-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31189-5G 31189-1G 31189-25G 31189-500MG
77632-638EA 841 USD
77632-638 77637-976 77654-920 77649-980
5-Chloro-2-fluoro-4-methoxyphenylboronic acid pinacol ester ≥97%
5-Chloro-2-fluoro-4-methoxyphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 286.535 g/mol
MW: C₁₃H₁₇BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD22414638
CAS Number: 2121513-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR