3-Chloro-6-fluoro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

31177-5G 31177-25G 31177-1G 31177-500MG
77634-716EA 218 USD
77634-716 77654-914 77641-938 77895-606
3-Chloro-6-fluoro-2-methoxybenzoic acid ≥97%
3-Chloro-6-fluoro-2-methoxybenzoic acid

Precaución: For Research Use Only

Formula: C₈H₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD22415170
CAS Number: 1781848-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR