Methyl 5-bromo-4-chloronicotinate ≥95%

Supplier: AOBChem USA

Warning

31167-5G 31167-1G 31167-25G 31167-500MG
77634-714EA 274 USD
77634-714 77637-972 77654-910 77649-972
Methyl 5-bromo-4-chloronicotinate ≥95%
Methyl 5-bromo-4-chloronicotinate

Precaución: For Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257863
CAS Number: 1256808-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR