5-Iodo-2,2-dimethyl-1,3-benzodioxole ≥97%

Supplier: AOBChem USA

Warning

311550-5G 311550-25G 311550-1G 311550-500MG
77632-632EA 1144 USD
77632-632 77658-626 77641-932 77645-946
5-Iodo-2,2-dimethyl-1,3-benzodioxole ≥97%
5-Iodo-2,2-dimethyl-1,3-benzodioxole

Precaución: For Research Use Only

Formula: 276.07 g/mol
MW: C₉H₉IO₂
Storage Temperature: Ambient
MDL Number: MFCD27922690
CAS Number: 861081-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR